Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WAY 161503 hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent and selective 5-HT2C receptor agonist (Ki = 4 nM; EC50 = 12 nM). Antidepressant following systemic administration in vivo.
Specifications
Quantity | 10 mg |
Synonym | way 161503 hydrochloride, 8,9-dichloro-2,3,4,4a-tetrahydro-1h-pyrazino 1,2-a quinoxalin-5 6h-one hydrochloride, 8,9-dichloro-1h,2h,3h,4h,4ah,6h-pyrazino 1,2-a quinoxalin-5-one hydrochloride, way161503 hydrochloride, 8,9-dichloro-2,3,4,4a-tetrahydro-1h,6h-pyrazino 1,2-a quinoxalin-5-one hydrochloride, way 161503 hydrochloride hplc, 1h-pyrazino 1,2-a quinoxalin-5 6h-one, 8,9-dichloro-2,3,4,4a-tetrahydro, 8,9-dichloro-1h,2h,3h,4h,4ah,6h-piperazino 1,2-a quinoxalin-5-one hydrochloride, 8,9-dichloro-2,3,4,4a-tetrahydro-1h-pyrazino 1,2-a quinoxalin-5 6h-one-hydrogen chloride 1/1 |
Formula Weight | 308.59 |
Percent Purity | >99% |
Chemical Name or Material | WAY 161503 hydrochloride |
Safety and Handling
Recommended Storage | Desiccate at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction