Learn More
WAY 200070, Tocris
Selective ERβ agonist
Manufacturer:  R&D Systems 3366/10
Quantity 10mg
 View more versions of this product
Catalog No. 33-661-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>98%
306.11
WAY 200070
C13H8BrNO3
7-Bromo-2-(4-hydroxyphenyl)-1, 3-benzoxazol-5-ol
Specifications
440122-66-7
10mg
306.11
WAY 200070
>98%
C13H8BrNO3
7-Bromo-2-(4-hydroxyphenyl)-1, 3-benzoxazol-5-ol
Safety and Handling
Recommended Storage : Store at Room Temperature