Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WBP11 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP258142

Catalog No. NBP258142


Add to cart

Description

Description

WBP11 Polyclonal antibody specifically detects WBP11 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

WBP11
Unconjugated
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
DKFZp779M1063, Npw38-binding protein, Npw38-binding protein NpwBP, NpwBP, NPWBPsplicing factor, PQBP1 and PP1 interacting, SH3 domain-binding protein SNP70, SIPP1WW domain-binding protein 11, SNP70, Splicing factor that interacts with PQBP-1 and PP1, WBP-11, WW domain binding protein 11
Rabbit
IgG
100 ul
Core ESC Like Genes, Stem Cell Markers
Primary
51729
Human
Western Blot, Immunocytochemistry, Immunofluorescence
Western Blot 0.4 ug/ml, Immunocytochemistry/Immunofluorescence 1-4 ug/ml
Affinity Purified
WBP11
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:DIYKELRKLEVEYEQKRAQLSQYFDAVKNAQHVEVESIPLPDMPHAPSNILIQDIPLPGAQPPSILKKTSAYGPPTRAVSILPLLGHGVPRLP
Affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only