Learn More
Aviva Systems Biology Corp WBP2 Antibody - N-terminal region (ARP54911_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP54911_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WBP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human WBP2. Alternate names: MGC18269, WBP-2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-61365