Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WBP2NL Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548895
Description
May play a role in meotic resumption and pronuclear formation, mediated by a WW domain-signaling pathway during fertilization.
Specifications
WBP2NL | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q6ICG8 | |
WBP2NL | |
KLH conjugated synthetic peptide between 9-36 amino acids from the N-terminal region of human WBP2NL. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
WBP2NL | |
Postacrosomal sheath WW domain-binding protein, WW domain-binding protein 2-like, WBP2NL, PAWP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
164684 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction