Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WBP2NL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | WBP2NL |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP152897
|
Novus Biologicals
NBP152897 |
100 μL |
Each of 1 for $482.50
|
|
|||||
Description
WBP2NL Polyclonal specifically detects WBP2NL in Human samples. It is validated for Western Blot.Specifications
WBP2NL | |
Polyclonal | |
Rabbit | |
Human | |
MGC26816, PAWPFLJ26145, postacrosomal sheath WW domain-binding protein, WBP2 N-terminal like, WW domain-binding protein 2-like | |
WBP2NL | |
IgG | |
32 kDa |
Western Blot | |
Unconjugated | |
RUO | |
Q6ICG8 | |
164684 | |
Synthetic peptides corresponding to WBP2NL(WBP2 N-terminal like) The peptide sequence was selected from the middle region of WBP2NL. Peptide sequence GYGAPPLGYGAPPAGNEGPPAGYRASPAGSGARPHESTAAQAPENEASLP. | |
Primary |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title