Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WBSCR16 Rabbit anti-Hamster, Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548652

Catalog No. PA548652

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a protein containing regulator of chromosome condensation 1-like repeats. The encoded protein may function as a guanine nucleotide exchange factor. This gene is located in a region of chromosome 7 that is deleted in Williams-Beuren syndrome, a multisystem developmental disorder. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Jul 2013] Orthologs mouse all
Specifications

Specifications

WBSCR16
Polyclonal
PBS with 0.09% sodium azide
Q96I51
Williams-Beuren syndrome chromosomal region 16 protein, RCC1-like G exchanging factor-like protein, WBSCR16
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100768632, 81554
Western Blot
Unconjugated
RCC1L
Liquid
Rcc1l
KLH conjugated synthetic peptide between 15-44 amino acids from the N-terminal region of human WBS16.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Hamster, Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit