Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDFY3 Rabbit anti-Human, Janelia Fluor 646, Polyclonal,Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP103332JF646

Catalog No. NBP13332646


Add to cart

Description

Description

WDFY3 Polyclonal antibody specifically detects WDFY3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

WDFY3
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
ALFY, Autophagy-linked FYVE protein, KIAA0993MGC16461, WD repeat and FYVE domain containing 3, WD repeat and FYVE domain-containing protein 3, ZFYVE25
Rabbit
IgG
0.1mL
Primary
23001
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Affinity Purified
WDFY3
The immunogen recognized by this antibody maps to a region between residue 500 and 550 of human WD repeat and FYVE domain containing 3 using the numbering given in entry NP_055806.2 (GeneID 23001).
Immunogen affinity purified
Store at 4°C in the dark.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit