Learn More
Aviva Systems Biology Corp WDR1 Antibody - N-terminal region (ARP41304_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP41304_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WDR1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human WDR1. Alternate names: AIP1, NORI-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-62881