Learn More
WDR18 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 151651AP
Catalog No. 50-173-0034
Add to cart
Description
Description
WD repeat domain 18 (WDR18) is a member of the WD repeat protein family, which is involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. WD repeats are minimally conserved regions of approximately 40 amino acids that commonly form a tertiary propeller structure. WDR18 is a 432 amino acid protein that contains six WD repeats and is encoded by a gene located on human chromosome 19. WDR18 may play a role during development. The complex PELP1-TEX10-WDR18 is required for the maturation and nucleolar release of the large ribosomal subunit.
Specifications
Specifications
WDR18
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BV38
R32184_1, WD repeat domain 18, WDR18
Rabbit
IgG
150 μL
-20°C
Primary
57418
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
WDR18
Liquid
WDR18
WDR18 Fusion Protein Ag7282
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human