Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WDR37 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599108
Description
WDR37 Polyclonal Antibody for Western Blot, IHC (P), ELISA
WDR37 encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes.
Specifications
WDR37 | |
Polyclonal | |
Unconjugated | |
WDR37 | |
fj52e09; im:7144367; Kiaa0982; RP11-529L18.2; WD repeat domain 37; WD repeat-containing protein 37; Wdr37; wu:fj52e09 | |
Rabbit | |
Protein G | |
RUO | |
22884 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
3.5 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q9Y2I8 | |
WDR37 | |
Recombinant Human WD repeat-containing protein 37 protein (1-264AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction