Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WDR53 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | WDR53 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP156804
|
Novus Biologicals
NBP156804 |
100 μL |
Each for $436.00
|
|
NBP15680420
|
Novus Biologicals
NBP15680420UL |
20 μL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
WDR53 Polyclonal specifically detects WDR53 in Human samples. It is validated for Western Blot.Specifications
WDR53 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
G-beta repeat-containing protein, MGC12928, MGC64882, WD repeat domain 53, WD repeat-containing protein 53 | |
WDR53 | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
Q7Z5U6 | |
348793 | |
Synthetic peptides corresponding to WDR53(WD repeat domain 53) The peptide sequence was selected from the middle region of WDR53. Peptide sequence NLLASADDSGAIKILDLENKKVIRSLKRHSNICSSVAFRPQRPQSLVSCG. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title