Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WDR61 Antibody (OTI4C4), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274866F
Description
WDR61 Monoclonal specifically detects WDR61 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
WDR61 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
WDR61 | |
Recombinant protein fragment corresponding to amino acids 92-305 of human WDR61 (NP_079510) produced in E.coli. | |
0.1 mL | |
80349.0 | |
Store at 4°C in the dark. |
Immunohistochemistry | |
OTI4C4 | |
Immunohistochemistry | |
Meiotic recombination REC14 protein homolog, REC14, recombination protein REC14, SKI8, WD repeat domain 61, WD repeat-containing protein 61 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction