Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDR61 Mouse anti-Human, HRP, Clone: OTI4C4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP274866H

Catalog No. NBP274866H


Add to cart

Description

Description

WDR61 Monoclonal antibody specifically detects WDR61 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

WDR61
OTI4C4
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
WDR61
Recombinant protein fragment corresponding to amino acids 92-305 of human WDR61 (NP_079510) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Western Blot
HRP
PBS with No Preservative
Meiotic recombination REC14 protein homolog, REC14, recombination protein REC14, SKI8, WD repeat domain 61, WD repeat-containing protein 61
Mouse
IgG1
0.1 mL
Primary
80349.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit