Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WDR61 Mouse anti-Human, Clone: OTI4C4, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP274866
Description
WDR61 Monoclonal antibody specifically detects WDR61 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
WDR61 | |
OTI4C4 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
Meiotic recombination REC14 protein homolog, REC14, recombination protein REC14, SKI8, WD repeat domain 61, WD repeat-containing protein 61 | |
Mouse | |
IgG1 | |
Immunogen affinity purified | |
Store at -20°C. Avoid freeze-thaw cycles. | |
Monoclonal | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. |
Immunohistochemistry, Western Blot | |
LYOPH | |
Western Blot 1:2000, Immunohistochemistry 1:150 | |
Affinity Purified, Lyophilized | |
WDR61 | |
Human recombinant protein fragment corresponding to amino acids 92-305 of human WDR61(NP_079510) produced in E.coli. | |
33.4 kDa | |
100 μg | |
Primary | |
80349 | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title