Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDR74 Mouse anti-Human, Mouse, Rat, Canine, Monkey, FITC, Clone: OTI4B2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP274867F

Catalog No. NBP274867F


Add to cart

Description

Description

WDR74 Monoclonal antibody specifically detects WDR74 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Immunohistochemistry.
Specifications

Specifications

WDR74
OTI4B2
Immunohistochemistry
Affinity Purified, Conjugated
WDR74
Recombinant protein fragment corresponding to amino acids 114-385 of human WDR74 (NP_060563) produced in E.coli.
Immunogen affinity purified
Stem Cell Markers
Primary
54663.0
Immunohistochemistry
FITC
PBS with 0.05% Sodium Azide
FLJ10439, FLJ21730, NOP seven-associated protein 1, NSA1, WD repeat domain 74, WD repeat-containing protein 74
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Canine, Human, Monkey, Mouse, Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit