Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDR86 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548389

Catalog No. PA548389


Add to cart

Specifications

Specifications

WDR86
Polyclonal
PBS with 0.09% sodium azide
Q86TI4
WD repeat-containing protein 86, WDR86
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
349136
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
WDR86
Liquid
WDR86
KLH conjugated synthetic peptide between 286-316 amino acids from the C-terminal region of human WDR86.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit