Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WDR89 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548574
Description
Predicted to react with bovine based on sequence homology.
Specifications
WDR89 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q96FK6 | |
WDR89 | |
KLH conjugated synthetic peptide between 359-387 amino acids from the C-terminal region of human WDR89. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
WDR89 | |
WD repeat-containing protein 89, WDR89, C14orf150 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
112840 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction