Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDR9 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen WDR9
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP154801
SDP
View Documents
Novus Biologicals
NBP154801
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

WDR9 Polyclonal specifically detects WDR9 in Human samples. It is validated for Western Blot.
Specifications

Specifications

WDR9
Polyclonal
Rabbit
Q6P2D1
54014
Synthetic peptides corresponding to BRWD1(bromodomain and WD repeat domain containing 1) The peptide sequence was selected from the N terminal of BRWD1. Peptide sequence MAEPSSARRPVPLIESELYFLIARYLSAGPCRRAAQVLVQELEQYQLLPK.
Primary
Western Blot
Unconjugated
RUO
bromodomain and WD repeat domain containing 1, bromodomain and WD repeat-containing protein 1, C21orf107, chromosome 21 open reading frame 107, FLJ11315, FLJ43918, N143, transcriptional unit N143, WD repeat domain 9, WD repeat protein WDR9-form2, WD repeat-containing protein 9, WDR9
BRWD1
IgG
13 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.