Learn More
WDR92 Mouse anti-Human, Clone: OTI2F2, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF809522
Catalog No. 50-166-9556
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
WD40 repeats are a common structural module in eukaryotic proteins, and proteins containing WD40 domains have a wide range of functions, including signal transduction, cell cycle regulation, RNA splicing, and transcription. One such protein, WDR92, also known as monad, contains two WD40 repeats is widely expressed in human tissues, especially testis. Overexpression of this protein or its binding partner RNA polymerase II-associated protein 3 (RPAP3) potentiated apoptosis and caspase-3 activation induced by TNF-a and cycloheximide, suggesting that WDR92, together with RPAP3, may function as a novel modulator of apoptosis.
Specifications
Specifications
WDR92
Monoclonal
Unconjugated
WDR92
Lyophilized
WDR92
Full length human recombit protein of human WDR92 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI2F2
PBS with 8% trehalose and no preservative; pH 7.3
Q96MX6
WD repeat domain 92
Mouse
IgG2b
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
116143