Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDR93 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548220

Catalog No. PA548220

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

WDR93
Polyclonal
PBS with 0.09% sodium azide
Q6P2C0
WD repeat-containing protein 93, WDR93
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
56964
Flow Cytometry, Western Blot
Unconjugated
WDR93
Liquid
WDR93
KLH conjugated synthetic peptide between 572-600 amino acids from the C-terminal region of human WDR93.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit