Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WDSUB1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | WDSUB1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP155073
|
Novus Biologicals
NBP155073 |
100 μL |
Each of 1 for $482.50
|
|
|||||
Description
WDSUB1 Polyclonal specifically detects WDSUB1 in Human samples. It is validated for Western Blot.Specifications
WDSUB1 | |
Polyclonal | |
Rabbit | |
Q8N9V3 | |
151525 | |
IgG | |
53 kDa |
Western Blot | |
Unconjugated | |
RUO | |
FLJ36175,2610014F08Rik, SAM and U-box domain-containing protein 1, UBOX6, WD repeat, SAM and U-box domain containing 1, WD repeat, sterile alpha motif and U-box domain containing 1, WDSAM1 | |
WDSUB1 | |
Primary |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title