Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wearwell™ Ortho Stand™ Mats
Domed anti fatigue mat increases circulation
Supplier: Wearwell™ 454.12X3X4BYL
View more versions of this product
Safety Services Available
Overview
Matting Assessments
Description
- An economical, dry area, single work area mat is ergonomically designed to increase worker comfort
- Hollow domed surface encourages subtle changes in posture increasing circulation and lessening fatigue
- ideal for checkout stands, manifest stations, mailing areas and punch presses
Order Info
Overall thickness: 0.5 in. (1.3cm)
Specifications
91.4 x 122 cm (3 x 4 ft.) | |
Black with Yellow Border | |
Natural Rubber |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction