Learn More
Wearwell™ Soft Rock Anti-fatigue Floor Mat
Modern, adaptable anti-fatigue matting solution for labs, hotels, retail outlets, and other facilities where workers stand on hard tiles or concrete
$81.10 - $218.50
Specifications
Thickness 1.3cm, 0.5 in.
Description Soft Rock
View More Specs
Products 12
Catalog Number Mfr. No. Type Color Price Quantity & Availability  
Catalog Number Mfr. No. Type Color Price Quantity & Availability  
19-000-018
View Documents
Wearwell™
423.12X2X3BL
Size: 61 x 91.4cm, 2 x 3 ft. Blue Stone Each for $90.00
Add to cart
19-000-019
View Documents
Wearwell™
423.12X2X3CL
Size: 61 x 91.4cm, 2 x 3 ft. Coal Each for $81.10
Add to cart
19-000-020
View Documents
Wearwell™
423.12X2X3GT
Size: 61 x 91.4cm, 2 x 3 ft. Granite Each for $90.00
Add to cart
19-000-021
View Documents
Wearwell™
423.12X2X3SA
Size: 61 x 91.4cm, 2 x 3 ft. Sandstone Each for $90.00
Add to cart
19-000-022
View Documents
Wearwell™
423.12X2X3SL
Size: 61 x 91.4cm, 2 x 3 ft. Slate Each for $90.00
Add to cart
19-000-023
View Documents
Wearwell™
423.12X2X3TR
Size: 61 x 91.4cm, 2 x 3 ft. Travertine Each for $90.00
Add to cart
19-000-024
View Documents
Wearwell™
423.12X3X5BL
Size: 91.4 x 152.4cm, 3 x 5 ft. Blue Stone Each for $218.50
Add to cart
19-000-025
View Documents
Wearwell™
423.12X3X5CL
Size: 91.4 x 152.4cm, 3 x 5 ft. Coal Each for $194.00
Add to cart
19-000-026
View Documents
Wearwell™
423.12X3X5GT
Size: 91.4 x 152.4cm, 3 x 5 ft. Granite Each for $218.50
Add to cart
19-000-027
View Documents
Wearwell™
423.12X3X5SA
Size: 91.4 x 152.4cm, 3 x 5 ft. Sandstone Each for $218.50
Add to cart
19-000-028
View Documents
Wearwell™
423.12X3X5SL
Size: 91.4 x 152.4cm, 3 x 5 ft. Slate Each for $218.50
Add to cart
19-000-029
View Documents
Wearwell™
423.12X3X5TR
Size: 91.4 x 152.4cm, 3 x 5 ft. Travertine Each for $218.50
Add to cart
Description
Description
  • Anti-microbial“Best”sponge
  • Abrasion and chemical resistant PVC surface
  • Machine cut, beveled edges prevent tripping
  • Slightly textured, easily cleaned surface
  • Variety of colors
  • Overall thickness of 0.5 in.
  • Suited for dry environments
Compliance
ASTM G21-13
Warranty and Services
4 years
Specifications
Specifications
1.3cm, 0.5 in.
Soft Rock