Learn More
Wearwell™ Mat Modular Diamond-Plate ESD Mat
Ideal for areas where static discharge is problem from microprocessor assembly, to general manufacturing areas
Manufacturer:  Wearwell™ 788.12X3X3GRBK
Catalog No. 19-041-192
Add to cart
Description
Description
  • The modular 3 x 3 ft. size is great for single workstations, but quickly connects to cover very large areas
  • Designed for comfort, they are also light-weight, easy to clean, and have rounded edges so additional bevels are not necessary
  • RTG = 3 x 104 to 5 x 105Ω
  • Mat must be grounded and employees must either wear heel grounders or conductive shoes
  • 0.5 in. thick
  • > 33% post industrial content
Specifications
Specifications
Natural Rubber
Solid diamond-plate surface