Learn More
Wearwell™ Mat Worksafe Light ESD Mat
Modular ESD mats are ideal for environments where static discharge is problem
Manufacturer: Wearwell™ 789.12X3X3GRBK
Description
- Use where airflow is important or debris is present
- The 3 x 3 ft. size is perfect for individual workstations, or interlocked to cover large areas
- Designed for comfort, it greatly reduces standing worker fatigue
- All edges are rounded for safety and each mat has grounding snap in one corner
- This mat must be grounded and employees must either wear heel grounders or conductive shoes
- 0.5 in. thick, lightweight and easy to clean
- Black
Specifications
Grease-resistant | |
Natural Rubber |