Learn More
Bel-Art™ SP Scienceware™ Weatherball Barometer Products for Science Education
Accurately predicts weather conditions.
Manufacturer:  Bel-Art™ B615010000
Catalog No. S95623
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Demonstrates impending weather changes to students
  • Rising water in the tube means bad weather; falling water in the tube means good weather
  • Includes four food-coloring tablets for better viewing of the changing weather conditions
Specifications
Specifications
Accurately predicts weather conditions.
Four food-coloring tablets
Weather Ball Barometer