Thermo Scientific™ WebSeal™ 96 Well Mats for use with WebSeal™ Kits
Extend the usefulness of your well plate kits with replacement mats.
Manufacturer: Thermo Scientific™ 60180M156
 View more versions of this product
Catalog No. 60-105-412
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Plate Compatibility WebSeal&trade Glass Inserted 96-Well Plates
    Type 96-Well WebSeal Mat, welled plugs
    Description 96-Well WebSeal Mat, welled plugs
    For Use With WebSeal Glass Inserted Plate Kits (60180-K108)
    Well Shape Welled plugs
    Pre-slit Yes
    No. per Pack 1
    Material Silicone/PTFE
    No. of Wells 96
    Extend the usefulness of your wellplate kits with Thermo Scientific™ WebSeal™ 96 Well Mats, which serve as accessories for WebSeal Glass Inserted Plate Kits.
  • Description & Specifications
Related Products
Product Content Feedback