Learn More
Thermo Scientific™ WebSeal™ 96-Well Plate Kits with Sealing Mats: PROMO
Save time and minimize the risk of cross-contamination with Thermo Scientific™ WebSeal™ 96-Well Plate Kits with Sealing Mats. Compatible with all 96-well-configured autosamplers, the ready-to-use kits consist of a comprehensive choice of 96-well microplates, pre-inserted glass or PTFE vials and silicone/PTFE sealing mats.
Products 8
Catalog Number Mfr. No. Description Microplate Vial Material Volume (Metric) Total Material Price Quantity & Availability  
Catalog Number Mfr. No. Description Microplate Vial Material Volume (Metric) Total Material Price Quantity & Availability  
03252191PM View Documents Thermo Scientific™
60180K100PROMO
96 Well Small Volume Microplate with Sealing Mat Glass Silicone/PTFE This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
03252192PM View Documents Thermo Scientific™
60180K101PROMO
96 Well Small Volume Microplate with Sealing Mat and Cutting Tool Glass Silicone/PTFE This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
03252193PM View Documents Thermo Scientific™
60180K103PROMO
96 Well Small Volume Microplate with Sealing Mat and Cutting Tool PTFE Silicone/PTFE This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
03252194PM View Documents Thermo Scientific™
60180K104PROMO
96 Well Microplate with Sealing Mat and Cutting Tool Glass Silicone/PTFE This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
03252195PM View Documents Thermo Scientific™
60180K105PROMO
96 Well Microplate with Crimp Top Vials, pre-inserted Glass Glass This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
03252196PM View Documents Thermo Scientific™
60180K106PROMO
96 Deep Well Plate with Sealing Mat Glass Silicone/PTFE This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
03252197PM View Documents Thermo Scientific™
60180K107PROMO
96 Square Well Plate with Sealing Mat Glass Silicone/PTFE This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
03252198PM View Documents Thermo Scientific™
60180K108PROMO
96 Square Well Plate with Sealing Mat - Precut Glass Silicone/PTFE This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Convenient wellplate kits include pre-inserted glass or PTFE vials and 96-position sealing mats; some include cutting tool
  • Mats seal samples for storage
  • Plates are glass-inserted for greater inertness compared to pure PP plates
  • Unique cutting tool allows removal of individual samples with the seal in place, eliminating cross-contamination
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.