Learn More
Sklar InstrumentsSupplier Diversity Partner Webster Needle Holder, Sklar, 5-1/4 in., Blunt, Straight, Serrated, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 201350
Webster Needle Holder, Sklar, 5-1/4 in., Blunt, Straight, Serrated, Stainless Steel. Sklar's Webster Needle Holder is used with small needles and fine suture material, commonly in plastic surgery or subcutaneous procedures. They are available with smooth or serrated jaws. Smooth jaws will cause the least damage to the needle; however, serrated jaws hold the needle more tightly. Physician preference typically determines the type of working surface utilized. This product is straight with smooth tips and a length of 5-1/4 inches.
Catalog No. 50-143-7451