Learn More
Sklar InstrumentsSupplier Diversity Partner TC Webster Needle Holder, Sklar, 5-1/4 in., Blunt, Straight, Cross-Serrated, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 218080
TC Webster Needle Holder, Sklar, 5-1/4 in., Blunt, Straight, Cross-Serrated, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. Sklar's TC Webster Needle Holder is a ratcheted, finger ring instrument used with small needles and fine suture material, commonly in plastic surgery or subcutaneous procedures. They are available with smooth or serrated jaws. Smooth jaws will cause the least damage to the needle, however, serrated jaws hold the needle more tightly. Physician preference typically determines the type of working surface utilized. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. This product is straight with cross-serrated tips and a length of 5-1/4 inches.
Catalog No. 50-143-7764
Due to product restrictions, please to purchase or view availability for this product.