Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Weighted Equivalency Sets
SDP

Perfect for use with pan balances in class demonstration or small-group activities.

Supplier:  Learning Resources, Inc LER3020

 View more versions of this product

Catalog No. S90839A

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Includes:
  • 24 number pieces
  • Activity guides

Description

Description

  • Make the relationship between fractions, decimals, and percents easy to understand
  • Colorful, plastic numbers are weighted, relative to the whole (100%, 50%, 25%, etc.)
  • Sets are interchangeable
  • Color coding aligns with the Rainbow Fraction™ System
  • Each number measures approximately 3 × 1 1/2 in.

Order Info

Grades K-5

Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit