Learn More
Sklar InstrumentsSupplier Diversity Partner Weisenbach Forceps Holder, Sklar, 6 in., Stainless Steel. Sklar's Weisenbach Forceps Holder has a t-bar design that allows one finger ring of the forceps handle to slide down the bar. This product is 6 inches. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 101775
Weisenbach Forceps Holder, Sklar, 6 in., Stainless Steel. Sklar's Weisenbach Forceps Holder has a t-bar design that allows one finger ring of the forceps handle to slide down the bar, thus separating it from the rest of the instrument for proper care and cleaning. The holder is availiable in 2 lengths, 6 inches and 12 inches. This product is 6 inches.
Catalog No. 50-143-7131