Learn More
Welch™ Compact Aspiration/Filtration Vacuum Stations: Pipette, Receiver Kit and Filter
Accumulate waste liquids safe and easily. Aspiration systems prevent ingestion of fluids leading to vacuum pump failure.
Manufacturer:  Welch™ 1475K21
 View more versions of this product
Catalog No. 01-258-121
  Check Availability
Add to cart
Includes: 40.5 oz. (1.2L) autoclaveable collection receiver, a hydrophobic inline filter, an automatic shutoff, vacuum regulator and gauge
Description
Description
  • Bleach resistant construction for extending service life
  • Continuous vacuum level monitoring with dial vacuum gauge
  • Powered by WOB-L piston vacuum pump
Recommended for aspirating aqueous and bleach solutions and not for acidic, basic or organic vapors and gases
Specifications
Specifications
Welch Compact Aspiration/Fitlration Vacuum Station
Replacement Hydrophobic Filter