Learn More
Welch™ Compact Aspiration/Filtration Vacuum Stations: Pipette, Receiver Kit and Filter
Accumulate waste liquids safe and easily. Aspiration systems prevent ingestion of fluids leading to vacuum pump failure.
Manufacturer:  Welch™ 1475K10
 View more versions of this product
Catalog No. 01-258-119
  Check Availability
Add to cart
Includes: 40.5 oz. (1.2L) autoclaveable collection receiver, a hydrophobic inline filter, an automatic shutoff, vacuum regulator and gauge
Description
Description
  • Bleach resistant construction for extending service life
  • Continuous vacuum level monitoring with dial vacuum gauge
  • Powered by WOB-L piston vacuum pump
Recommended for aspirating aqueous and bleach solutions and not for acidic, basic or organic vapors and gases
Specifications
Specifications
Stainless Steel aspiration adapter 4cm, Rubber adapter for Pasteur pipettes
Welch Compact Aspiration/Fitlration Vacuum Station
Hand Held Pipette