Learn More
Welch™ Oil-Mist Eliminator for Vacuum Pumps
Accessories for vacuum pumps
Manufacturer:  Welch™ 1416E01
 View more versions of this product
Catalog No. 01-108-1A
Add to cart
Description
Description
Oil-Mist Eliminator
  • Hermetically sealed
  • SS w/borosilicate microfiber element, flouroelastomer gaskets
  • Prevents corrosive exhaust gases from escaping
  • Accepts 13/16 in. hose
  • 12.7W x 19.68 in. H
Pump-Adapter Kit
  • For use with Oil-Mist Eliminator
Specifications
Specifications
Pump Adaptor Kit
Model 1400N vacuum pump