Learn More
Welch™ Oil for Vacuum Pumps
Accessory for direct drive DirecTorr vacuum pumps
Supplier:  Welch™ 8995P20
Catalog No. 01-131-28
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Low vapour pressure
  • Higher viscosity formula to resist breakdown at higher rpm and temperatures
  • No additives or inhibitors
  • 19L (5 gal.)
Specifications
Specifications
19 L
Direct drive DirecTorr™ vacuum pump
5 gal.
Vacuum Pump Oil