Learn More
Welch™ Replacement Gauge Tubes for Thermocouple Vacuum Gauges
Manufacturer:  Welch™ 1515A
Catalog No. 11-278-18
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
To assure maximum sensitivity over entire vacuum range
For vacuum gauge
0 to 5000μ
Thermocouple Gauge Sensor Tube