Learn More
Welch™ Economical High Capacity PTFE Diaphragm Vacuum Pumps
For oil-free chemical duty vacuum applications (1.5 to 760 TORR)
Supplier:  Welch™ 2064B01
 View more versions of this product
Catalog No. 12-584-503
Add to Cart
Description
Description
Long life and quiet operation
  • Handles corrosive gases and vapors
  • High flow, continous duty
  • For high vapor flow in larger scale laboratory vacuum applications
  • Used where vacuum pressure requirements are to the low Torr range(∽1.5 to 56 TORR) and low maintenance is a priority
  • All internal parts in contact with gases are made of chemically resistant fluoroplastics
  • Superior vapor tolerance and gas ballast valve for handling heavy vapor loads. The gas ballast valve preserves the pump performance under heavy vapor load by leaking air into second stage to inhibit condensation
  • Wired 115V, 60Hz with line cord and plug
Large vacuum ovens(>5 CF), floor model rotovaps, concentrators, pilot plants, vacuum source for filtration at multiple workstations, and vacuum autoclaves
Specifications
Specifications
Line cord and plug
Fluoroplastic
72.3 lb.
173 L/min.
60 Hz
115 V
115 V 60 Hz
2064
32.7 kg
Large vacuum ovens(>5 CF), floor model rotovaps, concentrators, pilot plants, vacuum source for filtration at multiple workstations and vacuum autoclaves
6 torr
Product Suggestions
Product Suggestions