Learn More
3M™ Welding Helmet
3M™ Welding Helmet may be configured for use with either Adflo or GVP turbo units.
Supplier: 3M™ 7000052731
Description
- May be configured for use with either Adflo or GVP turbo units and Supplied Air
- Turbo and SA units sold separately
- 3M Catalog# L-901SG , 3M Catalog# L-905SG
Specifications
ANSI Z87.1-2010 | |
Headgear | |
Welding and Wideview Faceshield | |
1 Ea. |
Green | |
Standard | |
L-905SG | |
Clear Visor and Auto-Darkening Welding Lens |