Wells Lamont All Day Inspector Gloves
Manufacturer: Wells Lamont Industry Group 3111MDFPM/L
 View more versions of this product
Catalog No. 19-048-692
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Cleaning Procedure Laundered using a mild soap and Detergent. Use a warm Water wash with a temperature not to exceed 160°F, wash time should not be longer than 15 to 20 min.. Use a rinse cycle for the product in cold Water, tumble dry glove in a temp
    Finger Style Full
    Glove Cuff Color Gold
    For Use With (Application) To create a barrier for Latex, Rubber and Plastic gloves, used as a glove liner or alone
    Laboratory/Cleanroom Use Yes
    Material Nylon, PHA Fiber
    Size Medium, Large
  • Description & Specifications
Related Products
Product Content Feedback