Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wells Lamont™ Cut Reistant Gloves
Click to view available options
Size:
Medium
Large
X-Large
Hem Color:
Blue
Brown
Green
Specifications
Color | Gray, Yellow |
Cut Resistance Rating | ANSI A4 |
Material | Stainless Steel Fiber, Polyester, Leather |
Product Type | Cut-resistant Gloves |
Size | Medium |
For Use With (Application) | High performance metal working and general industrial applications |
Glove Cuff Color | Green |
Hem Color | Green |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction