Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Economy Fleece Lined Grain Cowhide Gloves

Fleece lining provides comfort and warmth in cold environments. This work glove keeps hands warm in applications such as warehousing, and construction.

$2.25 - $10.25

Specifications

Color Natural
Glove Cuff Style Open
Grade Economy
Liner Material Fleece
Material Grain Cowhide Leather
View More Specs

Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-148-828
View Documents
Wells Lamont™
Y0062S
Small
Pair for $9.45
Pack of 12 PR for $113.40
Add to cart
 
19-148-827
View Documents
Wells Lamont™
Y0062M
Medium
Pair for $2.25

Save 50% Reg : $4.51

Pack of 12 PR for $27.00

Save 76% Reg : $113.40

Add to cart
 
19-148-826
View Documents
Wells Lamont™
Y0062L
Large
Pair for $9.40
Pack of 12 PR for $112.86
Add to cart
 
19-148-825
View Documents
Wells Lamont™
Y0062XL
X-Large
Pair for $10.25
Pack of 12 PR for $123.00
Add to cart
 
Description

Description

  • Grain cowhide with fleece lining
  • Shirred elastic back
  • Keystone thumb
  • Bound hem
Specifications

Specifications

Natural
Economy
Grain Cowhide Leather
Y0062
Open
Fleece
Drivers Gloves
Keystone
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit