Learn More
Wells Lamont™ FlexTech™ Foam Nitrile Coated Synthetic Knit Nylon Gloves
Lightweight nylon gloves feature foam nitrile coating on the palm and fingers for protection from cuts and abrasions.
Manufacturer:  Wells Lamont™ Y9249/XS/DEL
 View more versions of this product
Catalog No. 19-048-760
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
  • Black lightweight synthetic knit shell is comfortable and flexible
  • Foam nitrile coated palm enhances grip and provides resistance to snags and abrasions
  • Ideal for dry and slightly oily applications
Specifications
Specifications
Black
Electronics Assembly, and Small Parts Handling
Nylon
X-Small
Nitrile foam
Specific
No Texture
Palm, Fingers
Knit
Y9249
ANSI 4
ANSI A1
13
Seamless Knit