Learn More
Wells Lamont™ FlexTech™ Foam Nitrile-Coated Nylon Cut Resistant Gloves
Lightweight nylon gloves feature foam nitrile coating on the palm and fingers for protection from cuts and abrasions.
Manufacturer:  Wells Lamont™ Y9249/LG
 View more versions of this product
Catalog No. 19-048-763
Add to cart
Description
Description
  • Black lightweight synthetic knit shell with foam nitrile palm coating
  • Foam nitrile palm enhances grip and provides resistance to snags and abrasions
  • Ideal for dry and slightly oily applications
Specifications
Specifications
Black
Electronics Assembly, and Small Parts Handling
Nylon
Large
Nitrile Foam
Specific
Palm, Fingers
Knit
Y9249
ANSI 4
ANSI A1
13