Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ FlexTech™ Foam Nitrile Coated Synthetic Knit Nylon Gloves

Catalog Number 19048761
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

FlexTech™ Foam Nitrile Coated Synthetic Knit Nylon Gloves are lightweight nylon gloves featuring foam nitrile coating on the palm and fingers for protection from cuts and abrasions.

  • Black lightweight synthetic knit shell is comfortable and flexible
  • Foam nitrile coated palm enhances grip and provides resistance to snags and abrasions
  • Ideal for dry and slightly oily applications
ANSI Abrasion Rating ANSI 4
Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Nitrile Foam
Color Black
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size Small
For Use With (Application) Electronics Assembly, and Small Parts Handling
Glove Cuff Style Knit
Series Y9249
Yarn Gauge 13
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit