Wells Lamont™ FlexTech™ Gloves with Foam Nitrile Palm

Ideal for dry or slightly oily applications

Manufacturer: Wells Lamont Industry Group Y9249/XXL

 View more versions of this product

Catalog No. 19-048-765

  Check Availability
Add to Cart

    Specifications

    Exterior Finish Seamless
    Glove Cuff Style Over edge
    Glove Cuff Color Navy blue
    For Use With (Application) dry and slightly Oily applications and provides excellent grip and resistance to snags and abrasions
    Material Nitrile
    Liner Material Nylon
    Color Black
    Cut Resistance Rating Level 1
    Size 2X-Large
    Coated Area Palm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit