Wells Lamont™ FlexTech™ Synthetic Knit Shell with Full NBR Coating and Nitrile Palm

Excellent for general assembly, automotive, painting, manufacturing and the handling of small, wet and oily parts

Manufacturer: Wells Lamont Industry Group Y9289M

 View more versions of this product

Catalog No. 17-200-329

  Check Availability
Add to Cart

    Specifications

    Exterior Finish Sandy Nitrile
    Glove Cuff Style Synthetic Knit
    Glove Cuff Color Green
    For Use With (Application) Excellent for general assembly, automotive, painting, manufacturing and the handling of small, wet and oily parts
    Material HPPE
    Size Medium
    Thickness 13g
    Thumb Style Regular
    Coated Area Full palm
    Color Blue/Black
    Cut Resistance Rating Level 1

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit