Learn More
Wells Lamont™ FlexTech™ Full NBR Coated Synthetic Knit Gloves with Nitrile Coated Palm
Lightweight, synthetic knit shell is double coated with a layer of nitrile rubber (NBR) for water-resistance, and an additional layer of nitrile coating on the palm and fingers for enhanced grip capabilities.
$108.40
Specifications
ANSI Abrasion Rating ANSI 4
Coating Material Nitrile
Color Blue
Cut Resistance Rating ANSI A1
Glove Cuff Style Knit
View More Specs
Products 6
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-200-327
View Documents
Wells Lamont™
Y9289XS
X-Small
Dozen of 12 for $108.40
Add to cart
17-200-328
View Documents
Wells Lamont™
Y9289S
Small
Dozen of 12 for $108.40
Add to cart
17-200-330
View Documents
Wells Lamont™
Y9289L
Large
Dozen of 12 for $108.40
Add to cart
17-200-331
View Documents
Wells Lamont™
Y9289XL
X-Large
Dozen of 12 for $108.40
Add to cart
17-200-332
View Documents
Wells Lamont™
Y9289XXL
2X-Large
Dozen of 12 for $108.40
Add to cart
17-200-329
View Documents
Wells Lamont™
Y9289M
Medium This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Lightweight, synthetic knit shell with double coating
  • Sandy nitrile coated palm and fingertips enhances grip
  • Full NBR coating provides an additional layer of protection against liquids
  • Gloves offer a balance of comfort and dexterity
Specifications
Specifications
ANSI 4
Blue
Knit
Y9289
Nitrile
ANSI A1
Nitrile Rubber (NBR)
13