Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Grain Cowhide Leather Drivers Gloves with Keystone Thumb

Catalog Number 19823914
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Grain Cowhide Leather Drivers Gloves are constructed from high quality materials to provide comfort and protection while on the job. A shirred elastic wrist prevents particles from entering the glove, while a keystone thumb adds movement and comfort.

  • Economy option
  • Elastic shirred back
  • Bound hem
  • Keystone thumb
Back Material Shirred Elastic
Color Natural
Grade Economy
Hand Specific
Material Grain Cowhide Leather
Product Type Driver Gloves
Size Small
For Use With (Application) Landscaping, Farming, Warehousing, Construction, Oil and Gas Industries
Glove Cuff Style Open
Hem Color Red
Series Y0131
Thumb Style Keystone
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit